CAS RN 1346007-05-3
IUPAC Name:
4,4,5,5-tetramethyl-2-{9,9'-spirobi[fluoren]-6-yl}-1,3,2-dioxaborolane
Synonyms:
4,4,5,5-tetramethyl-2-{9,9'-spirobi[fluoren]-6-yl}-1,3,2-dioxaborolane
Molecular Weight (g/mol):
442.37
Molecular Formula:
C31H27BO2
InChi Key:
FCQPLZQSAKVCFH-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1(C3=CC=CC=C3C3=CC=CC=C13)C1=CC=CC=C21
1
–
1
of
1
results
1
2-(9,9'-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™
CAS: 1346007-05-3 Molecular Formula: C31H27BO2 Molecular Weight (g/mol): 442.365 InChI Key: FCQPLZQSAKVCFH-UHFFFAOYSA-N Synonym: (9,9′-Spirobi[fluoren]-3-yl)boronic Acid Pinacol Ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9′-spirobi[fluorene] PubChem CID: 68211018 IUPAC Name: 4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-3-yl)-1,3,2-dioxaborolane SMILES: B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4(C5=CC=CC=C5C6=CC=CC=C64)C7=CC=CC=C73