missing translation for 'onlineSavingsMsg'
Learn More

2-(9,9'-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. P2562200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2562200MG 200 mg
P25621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2562200MG Supplier TCI America Supplier No. P2562200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1346007-05-3
Molecular Formula C31H27BO2
Molecular Weight (g/mol) 442.365
InChI Key FCQPLZQSAKVCFH-UHFFFAOYSA-N
Synonym (9,9′-Spirobi[fluoren]-3-yl)boronic Acid Pinacol Ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9′-spirobi[fluorene]
PubChem CID 68211018
IUPAC Name 4,4,5,5-tetramethyl-2-(9,9'-spirobi[fluorene]-3-yl)-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4(C5=CC=CC=C5C6=CC=CC=C64)C7=CC=CC=C73

Specifications

Melting Point 216°C
Color White-Yellow
Quantity 200 mg
Formula Weight 442.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9,9′-Spirobi[fluoren]-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane