missing translation for 'onlineSavingsMsg'
Learn More

2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Supplier:  TCI America D50331G

 View more versions of this product

Catalog No. D50331G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
170°C
C31H29BO2
9,9-Diphenyl-9H-fluorene-4-boronic Acid Pinacol Ester, 9,9-Diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluorene
B1(OC(C(O1)(C)C)(C)C)C2=C3C4=CC=CC=C4C(C3=CC=C2)(C5=CC=CC=C5)C6=CC=CC=C6
444.381
444.38
Crystalline Powder
1259280-37-9
White
1 g
WTFLXHPXCVWFEP-UHFFFAOYSA-N
2-(9,9-diphenylfluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
68636917
≥98.0% (HPLC,T)

Chemical Identifiers

1259280-37-9
444.381
9,9-Diphenyl-9H-fluorene-4-boronic Acid Pinacol Ester, 9,9-Diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluorene
2-(9,9-diphenylfluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
C31H29BO2
WTFLXHPXCVWFEP-UHFFFAOYSA-N
68636917
B1(OC(C(O1)(C)C)(C)C)C2=C3C4=CC=CC=C4C(C3=CC=C2)(C5=CC=CC=C5)C6=CC=CC=C6
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions