missing translation for 'onlineSavingsMsg'
Learn More

2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. p-7128622
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5033200MG 200 mg
D50331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5033200MG Supplier TCI America Supplier No. D5033200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1259280-37-9
Molecular Formula C31H29BO2
Molecular Weight (g/mol) 444.381
InChI Key WTFLXHPXCVWFEP-UHFFFAOYSA-N
Synonym 9,9-Diphenyl-9H-fluorene-4-boronic Acid Pinacol Ester, 9,9-Diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluorene
PubChem CID 68636917
IUPAC Name 2-(9,9-diphenylfluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C3C4=CC=CC=C4C(C3=CC=C2)(C5=CC=CC=C5)C6=CC=CC=C6

Specifications

Melting Point 170°C
Color White
Quantity 200 mg
Formula Weight 444.38
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9,9-Diphenyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane