missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(9,9-Diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 97.0+%, TCI America™
Supplier: TCI America D5009200MG
Specifications
| 2-(9,9-Diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 200°C | |
| C31H29BO2 | |
| 200 mg | |
| CQMUDHYVDPNPLZ-UHFFFAOYSA-N | |
| 2-(9,9-diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
| 12014049 | |
| ≥97.0% (HPLC,T) |
| 462128-39-8 | |
| White | |
| MFCD29089356 | |
| (9,9-Diphenyl-9H-fluoren-2-yl)boronic Acid Pinacol Ester, 9,9-Diphenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluorene | |
| CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=CC=CC=C1C2(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 444.38 | |
| 444.38 | |
| Crystalline Powder |
Chemical Identifiers
| 462128-39-8 | |
| 444.38 | |
| CQMUDHYVDPNPLZ-UHFFFAOYSA-N | |
| 12014049 | |
| CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=CC=CC=C1C2(C1=CC=CC=C1)C1=CC=CC=C1 |
| C31H29BO2 | |
| MFCD29089356 | |
| (9,9-Diphenyl-9H-fluoren-2-yl)boronic Acid Pinacol Ester, 9,9-Diphenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluorene | |
| 2-(9,9-diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Safety and Handling
TSCA : No