missing translation for 'onlineSavingsMsg'
Learn More

2-(9,9-Dimethyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. p-7128611
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50181G 1 g
D50185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50181G Supplier TCI America Supplier No. D50181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 569343-09-5
Molecular Formula C21H25BO2
Molecular Weight (g/mol) 320.239
MDL Number MFCD08704229
InChI Key DAZFRJAIIUPRQZ-UHFFFAOYSA-N
Synonym 2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dimethylfluorene-2-boronic acid pinacol ester, 1,3,2-dioxaborolane, 2-9,9-dimethyl-9h-fluoren-2-yl-4,4,5,5-tetramethyl, 2-9,9-dimethylfluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, fl-be, amtb252, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dimethylfluorene, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborole-2-yl-9,9-dimethyl-9h-fluorene
PubChem CID 45358694
IUPAC Name 2-(9,9-dimethylfluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=CC=CC=C4C3(C)C

Specifications

Melting Point 136°C
Color White
Quantity 1 g
Formula Weight 320.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(9,9-Dimethyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane