missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ 2.8 M Tris HCl
GREENER_CHOICE

Catalog No. A65689BA
Change view
Click to view available options
Quantity:
5 L
20 L
50 L
100 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A65689BA 5 L
A65689BB 20 L
A65689BC 50 L
A65689BD 100 L
A65689BE 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. A65689BA Supplier Gibco™ Supplier No. A65689BA

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.

Gibco concentrates are intended for dilution at the point of use and come sterile-filtered and packaged in conveniently sized bioprocess containers. Sourcing concentrated stock solutions can help streamline buffer preparation by helping reduce preparation time, storage space, and the manufacturing footprint required for hydration.

Features

  • Concentrated for dilution on-site
  • Available in a range of pack sizes
  • Established shelf life: set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification

Benefits

  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages

Additional information

  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: TAegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard

Specifications

Form Clear, colorless solution
Product Type Tris-HCl
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Formulation Tris Buffer Component (Acidic)
Manufacturing Quality CGMP
Quantity 5 L

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.