missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,8-Diiododibenzothiophene 97.0+%, TCI America™
$251.65 - $870.51
Chemical Identifiers
| CAS | 105404-91-9 |
|---|---|
| Molecular Formula | C12H6I2S |
| Molecular Weight (g/mol) | 436.049 |
| InChI Key | JUMCSMONVKDUIB-UHFFFAOYSA-N |
| PubChem CID | 13530940 |
| IUPAC Name | 2,8-diiododibenzothiophene |
| SMILES | C1=CC2=C(C=C1I)C3=C(S2)C=CC(=C3)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4870200MG
|
TCI America
D4870200MG |
200 mg |
Each for $251.65
|
|
|||||
|
D48701G
|
TCI America
D48701G |
1 g |
Each for $870.51
|
|
|||||
Chemical Identifiers
| 105404-91-9 | |
| 436.049 | |
| 13530940 | |
| C1=CC2=C(C=C1I)C3=C(S2)C=CC(=C3)I |
| C12H6I2S | |
| JUMCSMONVKDUIB-UHFFFAOYSA-N | |
| 2,8-diiododibenzothiophene |
Specifications
| 105404-91-9 | |
| White-Yellow | |
| 200 mg | |
| C1=CC2=C(C=C1I)C3=C(S2)C=CC(=C3)I | |
| 436.049 | |
| 436.05 | |
| Crystalline Powder |
| 225°C | |
| C12H6I2S | |
| JUMCSMONVKDUIB-UHFFFAOYSA-N | |
| 2,8-diiododibenzothiophene | |
| 13530940 | |
| ≥97.0% (GC) | |
| 2,8-Diiododibenzothiophene |
Safety and Handling
TSCA : No