missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,8-Diiododibenzothiophene 97.0+%, TCI America™
Supplier: TCI America D48701G
Specifications
| 2,8-Diiododibenzothiophene | |
| 225°C | |
| C12H6I2S | |
| JUMCSMONVKDUIB-UHFFFAOYSA-N | |
| 2,8-diiododibenzothiophene | |
| 13530940 | |
| ≥97.0% (GC) |
| 105404-91-9 | |
| White-Yellow | |
| 1 g | |
| C1=CC2=C(C=C1I)C3=C(S2)C=CC(=C3)I | |
| 436.049 | |
| 436.05 | |
| Crystalline Powder |
Chemical Identifiers
| 105404-91-9 | |
| 436.049 | |
| 13530940 | |
| C1=CC2=C(C=C1I)C3=C(S2)C=CC(=C3)I |
| C12H6I2S | |
| JUMCSMONVKDUIB-UHFFFAOYSA-N | |
| 2,8-diiododibenzothiophene |
Safety and Handling
TSCA : No