missing translation for 'onlineSavingsMsg'
Learn More

2,8-Dichlorocyclooctanone 98.0+%, TCI America™

Catalog No. D29101G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29101G Supplier TCI America Supplier No. D29101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108249-93-0
Molecular Formula C8H12Cl2O
Molecular Weight (g/mol) 195.083
MDL Number MFCD06797089
InChI Key COOPJLNDURVHCV-UHFFFAOYSA-N
PubChem CID 44630271
IUPAC Name 2,8-dichlorocyclooctan-1-one
SMILES C1CCC(C(=O)C(CC1)Cl)Cl

Specifications

Melting Point 43°C
Color White
Boiling Point 110°C
Quantity 1 g
Formula Weight 195.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,8-Dichlorocyclooctanone
TSCA No
Recommended Storage Refrigerator