accessibility menu, dialog, popup

UserName

CAS RN 108249-93-0

IUPAC Name: 2,8-dichlorocyclooctan-1-one
Synonyms: 2,8-dichlorocyclooctan-1-one
Molecular Weight (g/mol): 195.08
Molecular Formula: C8H12Cl2O
InChi Key: COOPJLNDURVHCV-UHFFFAOYNA-N
SMILES: ClC1CCCCCC(Cl)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results