missing translation for 'onlineSavingsMsg'
Learn More

2,8-Dibromodibenzothiophene 5,5-Dioxide 95.0+%, TCI America™

Catalog No. D5027200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5027200MG 200 mg
D50271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D5027200MG Supplier TCI America Supplier No. D5027200MG
Only null left

Chemical Identifiers

CAS 40307-15-1
Molecular Formula C12H6Br2O2S
Molecular Weight (g/mol) 374.046
MDL Number MFCD00093102
InChI Key ZFGCKZCEDNBNMV-UHFFFAOYSA-N
Synonym 2,8-Dibromodibenzothiophene Sulfone, 2,8-Dibromodibenzosulfolane
PubChem CID 3905994
IUPAC Name 2,8-dibromodibenzothiophene 5,5-dioxide
SMILES C1=CC2=C(C=C1Br)C3=C(S2(=O)=O)C=CC(=C3)Br

Specifications

Melting Point 361°C
Quantity 200 mg
Formula Weight 374.05
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,8-Dibromodibenzothiophene 5,5-Dioxide