CAS RN 40307-15-1
IUPAC Name:
4,12-dibromo-8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-8,8-dione
Synonyms:
4,12-dibromo-8λ⁶-thiatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene-8,8-dione
Molecular Weight (g/mol):
374.05
Molecular Formula:
C12H6Br2O2S
InChi Key:
ZFGCKZCEDNBNMV-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)S(=O)(=O)C1=C2C=C(Br)C=C1
1
–
1
of
1
results
1
2,8-Dibromodibenzothiophene 5,5-Dioxide 95.0+%, TCI America™
CAS: 40307-15-1 Molecular Formula: C12H6Br2O2S Molecular Weight (g/mol): 374.046 MDL Number: MFCD00093102 InChI Key: ZFGCKZCEDNBNMV-UHFFFAOYSA-N Synonym: 2,8-Dibromodibenzothiophene Sulfone, 2,8-Dibromodibenzosulfolane PubChem CID: 3905994 IUPAC Name: 2,8-dibromodibenzothiophene 5,5-dioxide SMILES: C1=CC2=C(C=C1Br)C3=C(S2(=O)=O)C=CC(=C3)Br