missing translation for 'onlineSavingsMsg'
Learn More

(2,7-Octadien-1-yl)succinic Anhydride, TCI America™

Catalog No. O024425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O024425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O024425G Supplier TCI America Supplier No. O024425G
Only null left

Chemical Identifiers

CAS 142826-45-7
Molecular Formula C12H16O3
Molecular Weight (g/mol) 208.257
MDL Number MFCD00191676
InChI Key XPIQJMUYUKAKNX-VOTSOKGWSA-N
Synonym 4,9-Decadiene-1,2-dicarboxylic Anhydride
PubChem CID 5367357
IUPAC Name 3-[(2E)-octa-2,7-dienyl]oxolane-2,5-dione
SMILES C=CCCCC=CCC1CC(=O)OC1=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 208.26
Physical Form Liquid
Chemical Name or Material (2,7-Octadien-1-yl)succinic Anhydride