accessibility menu, dialog, popup

UserName

H2COOO(E/Z)

CAS RN 142826-45-7

IUPAC Name: 3-(octa-2,7-dien-1-yl)oxolane-2,5-dione
Synonyms: 3-(octa-2,7-dien-1-yl)oxolane-2,5-dione
Molecular Weight (g/mol): 208.26
Molecular Formula: C12H16O3
InChi Key: XPIQJMUYUKAKNX-UHFFFAOYNA-N
SMILES: C=CCCCC=CCC1CC(=O)OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results