missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dinitro-9-fluorenone 98.0+%, TCI America™

Catalog No. p-7126710
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D139625G 25 g
D1396250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D139625G Supplier TCI America Supplier No. D139625G
Only null left

Chemical Identifiers

CAS 31551-45-8
Molecular Formula C13H6N2O5
Molecular Weight (g/mol) 270.20
MDL Number MFCD00001153
InChI Key HDVGAFBXTXDYIB-UHFFFAOYSA-N
Synonym 2,7-dinitro-9-fluorenone, 2,7-dinitrofluorenone, 9h-fluoren-9-one, 2,7-dinitro, 2,7-dinitro-9h-fluoren-9-one, fluoren-9-one, 2,7-dinitro, 9-fluorenone, 2,7-dinitro, ccris 2911, acmc-209hnh, maybridge1_004322, fluoren-9-one,7-dinitro
PubChem CID 35842
IUPAC Name 2,7-dinitro-9H-fluoren-9-one
SMILES [O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O

Specifications

Melting Point 298°C
Color Yellow
Quantity 25 g
Formula Weight 270.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dinitro-9-fluorenone
RTECSNumber LL9051000
TSCA Yes