missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dinitro-9-fluorenone, 97%

Catalog No. p-7052656
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5517606 5 g
AAH5517614 25 g
AAH5517622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5517606 Supplier Thermo Scientific Chemicals Supplier No. H5517606
Only null left

CAS: 31551-45-8 | C13H6N2O5 | 270.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 31551-45-8
Molecular Formula C13H6N2O5
Molecular Weight (g/mol) 270.20
MDL Number MFCD00001153
InChI Key HDVGAFBXTXDYIB-UHFFFAOYSA-N
Synonym 2,7-dinitro-9-fluorenone, 2,7-dinitrofluorenone, 9h-fluoren-9-one, 2,7-dinitro, 2,7-dinitro-9h-fluoren-9-one, fluoren-9-one, 2,7-dinitro, 9-fluorenone, 2,7-dinitro, ccris 2911, acmc-209hnh, maybridge1_004322, fluoren-9-one,7-dinitro
PubChem CID 35842
IUPAC Name 2,7-dinitrofluoren-9-one
SMILES [O-][N+](=O)C1=CC=C2C3=CC=C(C=C3C(=O)C2=C1)[N+]([O-])=O

Specifications

Melting Point 292°C to 295°C
Color Yellow to Orange
Quantity 5 g
Formula Weight 270.2
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2,7-Dinitro-9-fluorenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 250-695-1
RTECSNumber LL9051000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only