missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dimethyloctane, 98%, Thermo Scientific Chemicals

Catalog No. AAB2164603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2164603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2164603 Supplier Thermo Scientific Chemicals Supplier No. B2164603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-16-8
Molecular Formula C10H22
Molecular Weight (g/mol) 142.286
MDL Number MFCD00049100
InChI Key KEVMYFLMMDUPJE-UHFFFAOYSA-N
Synonym octane, 2,7-dimethyl, diisoamyl, 2,7dimethyloctane, diisopentyl, octane,7-dimethyl, 2,7-dimethyl-octane, acmc-1bq62
PubChem CID 14070
IUPAC Name 2,7-dimethyloctane
SMILES CC(C)CCCCC(C)C

Specifications

Melting Point -55°C
Density 0.72
Boiling Point 159°C to 160°C
Refractive Index 1.409
Linear Formula (CH3)2CH(CH2)4CH(CH3)2
Quantity 1 g
UN Number UN3295
Formula Weight 142.29
Percent Purity 98%
Chemical Name or Material 2,7-Dimethyloctane
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only