CAS RN 1072-16-8
IUPAC Name:
2,7-dimethyloctane
Synonyms:
octane, 2,7-dimethyl-
Molecular Weight (g/mol):
142.29
Molecular Formula:
C10H22
InChi Key:
KEVMYFLMMDUPJE-UHFFFAOYSA-N
SMILES:
CC(C)CCCCC(C)C
1
–
1
of
1
results
1
Promotions Available
2,7-Dimethyloctane, 98%, Thermo Scientific Chemicals
2,7-Dimethyloctane, 98%, Thermo Scientific Chemicals
CAS: 1072-16-8 Molecular Formula: C10H22 Molecular Weight (g/mol): 142.286 MDL Number: MFCD00049100 InChI Key: KEVMYFLMMDUPJE-UHFFFAOYSA-N Synonym: octane, 2,7-dimethyl,diisoamyl,2,7dimethyloctane,diisopentyl,octane,7-dimethyl,2,7-dimethyl-octane,acmc-1bq62 PubChem CID: 14070 IUPAC Name: 2,7-dimethyloctane SMILES: CC(C)CCCCC(C)C