missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 2',7'-Difluorofluorescein (Oregon Green™ 488)

Catalog No. D6145
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D6145 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D6145 Supplier Invitrogen™ Supplier No. D6145

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

2',7'-difluorofluorescein (Oregon Green™ 488)

Oregon Green™ 488 can be used as a reference standard for Oregon Green™ 488 conjugates or a pH sensor of moderately acidic solutions (pKa ∼4.7)

Specifications

Quantity 10 mg
Product Type 2',7'-Difluorofluorescein
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
Product Line Oregon Green

For Research Use Only. Not for use in diagnostic procedures.