missing translation for 'onlineSavingsMsg'
Learn More

2',7'-Dichlorofluorescein 3',6'-diacetate, 97%

Catalog No. p-105019
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC347250010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC347250010 Supplier Thermo Scientific Chemicals Supplier No. 347250010
Only null left

CAS: 2044-85-1 | C24H14Cl2O7 | 485.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2044-85-1
Molecular Formula C24H14Cl2O7
Molecular Weight (g/mol) 485.27
MDL Number MFCD00037501
InChI Key VQVUBYASAICPFU-UHFFFAOYSA-N
PubChem CID 104913
IUPAC Name (6'-acetyloxy-2',7'-dichloro-3-oxospiro[2-benzofuran-1,9'-xanthene]-3'-yl) acetate
SMILES CC(=O)OC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(OC(C)=O)C=C1O2

Specifications

Melting Point 239°C to 246°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 19, IV, 2912
Infrared Spectrum Authentic
Formula Weight 485.27
Percent Purity ≥96.0%
Packaging Glass bottle
Color Colorless to White
Quantity 1 g
Physical Form Powder or Crystals
Chemical Name or Material 2', 7'-Dichlorofluorescein 3', 6'-diacetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only