accessibility menu, dialog, popup

UserName

CH3OOOOH3COClOOCl

CAS RN 2044-85-1

IUPAC Name: 3'-(acetyloxy)-2',7'-dichloro-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-6'-yl acetate
Synonyms: 3'-(acetyloxy)-2',7'-dichloro-3-oxospiro[2-benzofuran-1,9'-xanthen]-6'-yl acetate
Molecular Weight (g/mol): 485.27
Molecular Formula: C24H14Cl2O7
InChi Key: VQVUBYASAICPFU-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC2=C(C=C1Cl)C1(OC(=O)C3=CC=CC=C13)C1=CC(Cl)=C(OC(C)=O)C=C1O2
Color 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)