missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dichlorofluorene, 97%, Thermo Scientific Chemicals

Catalog No. AAB2471706
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2471706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2471706 Supplier Thermo Scientific Chemicals Supplier No. B2471706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7012-16-0 | Molecular Formula: C13H8Cl2 | Molecular Weight: 235.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7012-16-0
Molecular Formula C13H8Cl2
Molecular Weight (g/mol) 235.107
MDL Number MFCD00032840
InChI Key SDPURBHAHVFTGX-UHFFFAOYSA-N
Synonym 2,7-dichlorofluorene, 2,7-dichlor-9h-fluoren, 9h-fluorene, 2,7-dichloro, 2,7-dichiorofluorene, fluorene,7-dichloro, 9h-fluorene,7-dichloro, ksc378s9n, 2,7-dichlorofluorene 1g
PubChem CID 251987
IUPAC Name 2,7-dichloro-9H-fluorene
SMILES C1C2=C(C=CC(=C2)Cl)C3=C1C=C(C=C3)Cl

Specifications

Melting Point >300°C (decomposition)
Quantity 5 g
Formula Weight 235.12
Percent Purity 97%
Chemical Name or Material 2,7-Dichlorofluorene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only