missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromopyrene 97.0+%, TCI America™

Catalog No. d31691g
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31691G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D31691G Supplier TCI America Supplier No. D31691G
Only null left

Chemical Identifiers

CAS 102587-98-4
Molecular Formula C16H8Br2
Molecular Weight (g/mol) 360.05
MDL Number MFCD09909862
InChI Key IGTQPXMEWQTTBJ-UHFFFAOYSA-N
PubChem CID 13615479
IUPAC Name 2,7-dibromopyrene
SMILES BrC1=CC2=CC=C3C=C(Br)C=C4C=CC(=C1)C2=C34

Specifications

Melting Point 321°C
Quantity 1 g
Formula Weight 360.05
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromopyrene