CAS RN 102587-98-4
IUPAC Name:
2,7-dibromopyrene
Synonyms:
2,7-dibromopyrene
Molecular Weight (g/mol):
360.05
Molecular Formula:
C16H8Br2
InChi Key:
IGTQPXMEWQTTBJ-UHFFFAOYSA-N
SMILES:
BrC1=CC2=CC=C3C=C(Br)C=C4C=CC(=C1)C2=C34
1
–
1
of
1
results
1
2,7-Dibromopyrene 97.0+%, TCI America™
CAS: 102587-98-4 Molecular Formula: C16H8Br2 Molecular Weight (g/mol): 360.05 MDL Number: MFCD09909862 InChI Key: IGTQPXMEWQTTBJ-UHFFFAOYSA-N PubChem CID: 13615479 IUPAC Name: 2,7-dibromopyrene SMILES: BrC1=CC2=CC=C3C=C(Br)C=C4C=CC(=C1)C2=C34