missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromophenanthrene-9,10-dione 97.0+%, TCI America™

Catalog No. D43631G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43631G 1 g
D43635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43631G Supplier TCI America Supplier No. D43631G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 84405-44-7
Molecular Formula C14H6Br2O2
Molecular Weight (g/mol) 366.01
MDL Number MFCD03931078
InChI Key LTZIIBSPYWTOQV-UHFFFAOYSA-N
PubChem CID 265843
IUPAC Name 2,7-dibromo-9,10-dihydrophenanthrene-9,10-dione
SMILES BrC1=CC=C2C3=CC=C(Br)C=C3C(=O)C(=O)C2=C1

Specifications

Melting Point 331°C
Quantity 1 g
Formula Weight 366.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromophenanthrene-9,10-dione