missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromofluorene 98.0+%, TCI America™

Catalog No. p-7127789
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35565G 5 g
D355625G 25 g
D3556100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D35565G Supplier TCI America Supplier No. D35565G
Only null left

Chemical Identifiers

CAS 16433-88-8
Molecular Formula C13H8Br2
Molecular Weight (g/mol) 324.02
MDL Number MFCD00019048
InChI Key AVXFJPFSWLMKSG-UHFFFAOYSA-N
PubChem CID 140073
IUPAC Name 2,7-dibromo-9H-fluorene
SMILES BrC1=CC=C2C(CC3=CC(Br)=CC=C23)=C1

Specifications

Melting Point 166°C
Color White-Yellow
Quantity 5 g
Synonym 2,7-dibromofluorene, 9h-fluorene, 2,7-dibromo, fluorene, 2,7-dibromo, fluorene,7-dibromo, 2, 7-dibromofluorene, 2,7-di bromo fluorene, 9h-fluorene,7-dibromo, acmc-1c8sn, ksc491i1j
Formula Weight 324.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromofluorene