missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromofluorene, 98%

Catalog No. p-7082146
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1243709 10 g
AAA1243714 25 g
AAA1243722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1243709 Supplier Thermo Scientific Chemicals Supplier No. A1243709

CAS: 16433-88-8 | C13H8Br2 | 324.02 g/mol

2,7-Dibromofluorene is used as a template for the N-carbazole capped oligofluorenes which show potential as hole-transporting materials for organic light emitting devices (OLEDs). It is also used in the synthesis of conjugated polymer, poly[9,9?-bis(6??-N,N,N-trimethylammonium)hexyl)fluorene-co-alt-4,7-(2,1,3-benzothiadiazole) dibromide] (PFBT), used in label-free DNA microarrays, in the preparation of blue photoluminescent unsymmetrically substituted polyfluorene and in the preparation of 2,7-dibromofluorene monomers containing benzyl ether dendrons (generations 1, 2 and 3) in the 9,9?-position of the fluorene ring, such as: 9,9-bis[(3,5-bis(benzyloxy)benzyloxy)methyl]-2,7-dibromofluorene, 9,9-bis[(3,5-bis(3,5-bis(benzyloxy)benzyloxy)benzyloxy)-methyl] 2,7-dibromo-fluorene, 9,9-bis[(3,5-bis(3,5-bis(3,5-bis(benzyloxy)benzyloxy)benzyloxy) benzyloxy)-methyl]-2,7-dibromofluorene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,7-Dibromofluorene is used as a template for the N-carbazole capped oligofluorenes which show potential as hole-transporting materials for organic light emitting devices (OLEDs). It is also used in the synthesis of conjugated polymer, poly[9,9′-bis(6′′-N,N,N-trimethylammonium)hexyl)fluorene-co-alt-4,7-(2,1,3-benzothiadiazole) dibromide] (PFBT), used in label-free DNA microarrays, in the preparation of blue photoluminescent unsymmetrically substituted polyfluorene and in the preparation of 2,7-dibromofluorene monomers containing benzyl ether dendrons (generations 1, 2 and 3) in the 9,9′-position of the fluorene ring, such as: 9,9-bis[(3,5-bis(benzyloxy)benzyloxy)methyl]-2,7-dibromofluorene, 9,9-bis[(3,5-bis(3,5-bis(benzyloxy)benzyloxy)benzyloxy)-methyl] 2,7-dibromo-fluorene, 9,9-bis[(3,5-bis(3,5-bis(3,5-bis(benzyloxy)benzyloxy)benzyloxy) benzyloxy)-methyl]-2,7-dibromofluorene.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 16433-88-8
Molecular Formula C13H8Br2
Molecular Weight (g/mol) 324.02
MDL Number MFCD00019048
InChI Key AVXFJPFSWLMKSG-UHFFFAOYSA-N
PubChem CID 140073
IUPAC Name 2,7-dibromo-9H-fluorene
SMILES BrC1=CC=C2C(CC3=CC(Br)=CC=C23)=C1

Specifications

Melting Point 164°C to 167°C
Quantity 10 g
Beilstein 2049205
Synonym 2,7-dibromofluorene, 9h-fluorene, 2,7-dibromo, fluorene, 2,7-dibromo, fluorene,7-dibromo, 2, 7-dibromofluorene, 2,7-di bromo fluorene, 9h-fluorene,7-dibromo, acmc-1c8sn, ksc491i1j
Formula Weight 324.03
Percent Purity 98%
Chemical Name or Material 2,7-Dibromofluorene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only