missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9-n-octylcarbazole 98.0+%, TCI America™

Catalog No. D3935200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3935200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3935200MG Supplier TCI America Supplier No. D3935200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 726169-75-1
Molecular Formula C20H23Br2N
Molecular Weight (g/mol) 437.22
MDL Number MFCD19440901
InChI Key GVRVHDUHAVNFOW-UHFFFAOYSA-N
PubChem CID 66979057
IUPAC Name 2,7-dibromo-9-octyl-9H-carbazole
SMILES CCCCCCCCN1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2

Specifications

Melting Point 71°C
Color Yellow
Quantity 200 mg
Formula Weight 437.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-n-octylcarbazole