accessibility menu, dialog, popup

UserName

H3CNBrBr

CAS RN 726169-75-1

IUPAC Name: 2,7-dibromo-9-octyl-9H-carbazole
Synonyms: 2,7-dibromo-9-octylcarbazole
Molecular Weight (g/mol): 437.22
Molecular Formula: C20H23Br2N
InChi Key: GVRVHDUHAVNFOW-UHFFFAOYSA-N
SMILES: CCCCCCCCN1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results