missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dibromo-9-fluorenone 98.0+%, TCI America™
$48.46 - $141.67
Chemical Identifiers
| CAS | 14348-75-5 |
|---|---|
| Molecular Formula | C13H6Br2O |
| Molecular Weight (g/mol) | 337.998 |
| MDL Number | MFCD00010790 |
| InChI Key | CWGRCRZFJOXQFV-UHFFFAOYSA-N |
| Synonym | 2,7-dibromo-9-fluorenone, 2,7-dibromo-9h-fluoren-9-one, 2,7-dibromo-fluoren-9-one, 2,7-dibromofluorenone, 2,7-dibromo-fluorenone, 9h-fluoren-9-one, 2,7-dibromo, 2,7-dibrom-9h-fluoren-9-on, pubchem19652, acmc-209cqp, maybridge1_002427 |
| PubChem CID | 259922 |
| IUPAC Name | 2,7-dibromofluoren-9-one |
| SMILES | C1=CC2=C(C=C1Br)C(=O)C3=C2C=CC(=C3)Br |
Chemical Identifiers
| 14348-75-5 | |
| 337.998 | |
| CWGRCRZFJOXQFV-UHFFFAOYSA-N | |
| 259922 | |
| C1=CC2=C(C=C1Br)C(=O)C3=C2C=CC(=C3)Br |
| C13H6Br2O | |
| MFCD00010790 | |
| 2,7-dibromo-9-fluorenone, 2,7-dibromo-9h-fluoren-9-one, 2,7-dibromo-fluoren-9-one, 2,7-dibromofluorenone, 2,7-dibromo-fluorenone, 9h-fluoren-9-one, 2,7-dibromo, 2,7-dibrom-9h-fluoren-9-on, pubchem19652, acmc-209cqp, maybridge1_002427 | |
| 2,7-dibromofluoren-9-one |
Specifications
| 14348-75-5 | |
| Yellow | |
| MFCD00010790 | |
| 2,7-dibromo-9-fluorenone, 2,7-dibromo-9h-fluoren-9-one, 2,7-dibromo-fluoren-9-one, 2,7-dibromofluorenone, 2,7-dibromo-fluorenone, 9h-fluoren-9-one, 2,7-dibromo, 2,7-dibrom-9h-fluoren-9-on, pubchem19652, acmc-209cqp, maybridge1_002427 | |
| C1=CC2=C(C=C1Br)C(=O)C3=C2C=CC(=C3)Br | |
| 337.998 | |
| 338.00 | |
| Crystalline Powder |
| 204°C | |
| C13H6Br2O | |
| 5 g | |
| CWGRCRZFJOXQFV-UHFFFAOYSA-N | |
| 2,7-dibromofluoren-9-one | |
| 259922 | |
| ≥98.0% (GC) | |
| 2,7-Dibromo-9-fluorenone |
Safety and Handling
TSCA : No