accessibility menu, dialog, popup

UserName

BrBrO

CAS RN 14348-75-5

IUPAC Name: 2,7-dibromo-9H-fluoren-9-one
Synonyms: 2,7-dibromofluoren-9-one
Molecular Weight (g/mol): 338
Molecular Formula: C13H6Br2O
InChi Key: CWGRCRZFJOXQFV-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C3=CC=C(Br)C=C3C(=O)C2=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)