missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9-fluorenone 98.0+%, TCI America™

Catalog No. D35575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35575G 5 g
D355725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35575G Supplier TCI America Supplier No. D35575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14348-75-5
Molecular Formula C13H6Br2O
Molecular Weight (g/mol) 337.998
MDL Number MFCD00010790
InChI Key CWGRCRZFJOXQFV-UHFFFAOYSA-N
Synonym 2,7-dibromo-9-fluorenone, 2,7-dibromo-9h-fluoren-9-one, 2,7-dibromo-fluoren-9-one, 2,7-dibromofluorenone, 2,7-dibromo-fluorenone, 9h-fluoren-9-one, 2,7-dibromo, 2,7-dibrom-9h-fluoren-9-on, pubchem19652, acmc-209cqp, maybridge1_002427
PubChem CID 259922
IUPAC Name 2,7-dibromofluoren-9-one
SMILES C1=CC2=C(C=C1Br)C(=O)C3=C2C=CC(=C3)Br

Specifications

Melting Point 204°C
Color Yellow
Quantity 5 g
Formula Weight 338.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-fluorenone