missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dibromo-9-(9-heptadecyl)carbazole 98.0+%, TCI America™
Supplier: TCI America D44901G
Specifications
| 2,7-Dibromo-9-(9-heptadecyl)carbazole | |
| 72°C | |
| C29H41Br2N | |
| 1 g | |
| CCCCCCCCC(CCCCCCCC)N1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2 | |
| 563.46 | |
| 563.46 | |
| Crystalline Powder |
| 955964-73-5 | |
| White-Yellow | |
| MFCD16619379 | |
| HRUNMSCVUSIWQM-UHFFFAOYSA-N | |
| 2,7-dibromo-9-(heptadecan-9-yl)-9H-carbazole | |
| 53400972 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 955964-73-5 | |
| 563.46 | |
| HRUNMSCVUSIWQM-UHFFFAOYSA-N | |
| 2,7-dibromo-9-(heptadecan-9-yl)-9H-carbazole |
| C29H41Br2N | |
| MFCD16619379 | |
| 53400972 | |
| CCCCCCCCC(CCCCCCCC)N1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2 |
Safety and Handling
TSCA : No