missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9-(9-heptadecyl)carbazole 98.0+%, TCI America™

Catalog No. p-7128303
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4490200MG 200 mg
D44901G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4490200MG Supplier TCI America Supplier No. D4490200MG
Only null left

Chemical Identifiers

CAS 955964-73-5
Molecular Formula C29H41Br2N
Molecular Weight (g/mol) 563.46
MDL Number MFCD16619379
InChI Key HRUNMSCVUSIWQM-UHFFFAOYSA-N
PubChem CID 53400972
IUPAC Name 2,7-dibromo-9-(heptadecan-9-yl)-9H-carbazole
SMILES CCCCCCCCC(CCCCCCCC)N1C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 200 mg
Formula Weight 563.46
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-(9-heptadecyl)carbazole