missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Dibromo-9,9-dihexylfluorene 98.0+%, TCI America™
$57.79 - $225.56
Chemical Identifiers
| CAS | 189367-54-2 |
|---|---|
| Molecular Formula | C25H32Br2 |
| Molecular Weight (g/mol) | 492.339 |
| MDL Number | MFCD03427215 |
| InChI Key | OXFFIMLCSVJMHA-UHFFFAOYSA-N |
| Synonym | 9,9-dihexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene, 2,7-dibromo-9,9-di-n-hexylfluorene, 9h-fluorene, 2,7-dibromo-9,9-dihexyl, 2,7-dibromo-9,9-dihexyl-fluorene, acmc-1c0sw, ksc491i1r, 9,9-di-n-hexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene homopolymer |
| PubChem CID | 3539647 |
| IUPAC Name | 2,7-dibromo-9,9-dihexylfluorene |
| SMILES | CCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCC |
Chemical Identifiers
| 189367-54-2 | |
| 492.339 | |
| OXFFIMLCSVJMHA-UHFFFAOYSA-N | |
| 3539647 | |
| CCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCC |
| C25H32Br2 | |
| MFCD03427215 | |
| 9,9-dihexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene, 2,7-dibromo-9,9-di-n-hexylfluorene, 9h-fluorene, 2,7-dibromo-9,9-dihexyl, 2,7-dibromo-9,9-dihexyl-fluorene, acmc-1c0sw, ksc491i1r, 9,9-di-n-hexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene homopolymer | |
| 2,7-dibromo-9,9-dihexylfluorene |
Specifications
| 189367-54-2 | |
| White-Yellow | |
| MFCD03427215 | |
| 9,9-dihexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene, 2,7-dibromo-9,9-di-n-hexylfluorene, 9h-fluorene, 2,7-dibromo-9,9-dihexyl, 2,7-dibromo-9,9-dihexyl-fluorene, acmc-1c0sw, ksc491i1r, 9,9-di-n-hexyl-2,7-dibromofluorene, 2,7-dibromo-9,9-dihexyl-9h-fluorene homopolymer | |
| CCCCCCC1(C2=C(C=CC(=C2)Br)C3=C1C=C(C=C3)Br)CCCCCC | |
| 492.339 | |
| 492.34 | |
| Crystalline Powder |
| 68°C | |
| C25H32Br2 | |
| 5 g | |
| OXFFIMLCSVJMHA-UHFFFAOYSA-N | |
| 2,7-dibromo-9,9-dihexylfluorene | |
| 3539647 | |
| ≥98.0% (HPLC) | |
| 2,7-Dibromo-9,9-dihexylfluorene |
Safety and Handling
TSCA : No