missing translation for 'onlineSavingsMsg'
Learn More

2,7-Dibromo-9,9-bis[3-(dimethylamino)propyl]fluorene 98.0+%, TCI America™

Catalog No. p-7128335
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4550200MG 200 mg
D45501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4550200MG Supplier TCI America Supplier No. D4550200MG

Chemical Identifiers

CAS 673474-73-2
Molecular Formula C23H30Br2N2
Molecular Weight (g/mol) 494.32
MDL Number MFCD22988888
InChI Key RJIWYGUYDXBQCJ-UHFFFAOYSA-N
PubChem CID 71721491
IUPAC Name (3-{2,7-dibromo-9-[3-(dimethylamino)propyl]-9H-fluoren-9-yl}propyl)dimethylamine
SMILES CN(C)CCCC1(CCCN(C)C)C2=C(C=CC(Br)=C2)C2=C1C=C(Br)C=C2

Specifications

Melting Point 76°C
Quantity 200 mg
Formula Weight 494.32
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material 2,7-Dibromo-9,9-bis[3-(dimethylamino)propyl]fluorene