missing translation for 'onlineSavingsMsg'
Learn More

2,7-Diamino-9,9-di-n-octylfluorene 98.0+%, TCI America™

Catalog No. p-7128422
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4719200MG 200 mg
D47191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4719200MG Supplier TCI America Supplier No. D4719200MG
Only null left

Chemical Identifiers

CAS 851042-10-9
Molecular Formula C29H44N2
Molecular Weight (g/mol) 420.69
MDL Number MFCD27923070
InChI Key ODJBGRHFVFWTBO-UHFFFAOYSA-N
PubChem CID 59306005
IUPAC Name 9,9-dioctyl-9H-fluorene-2,7-diamine
SMILES CCCCCCCCC1(CCCCCCCC)C2=C(C=CC(N)=C2)C2=C1C=C(N)C=C2

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 420.69
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Diamino-9,9-di-n-octylfluorene