missing translation for 'onlineSavingsMsg'
Learn More

2,7-Di-tert-butylnaphthalene 98.0+%, TCI America™

Catalog No. D23701G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23701G Supplier TCI America Supplier No. D23701G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10275-58-8
Molecular Formula C18H24
Molecular Weight (g/mol) 240.39
MDL Number MFCD00094397
InChI Key IHRUNSCACKKOPP-UHFFFAOYSA-N
PubChem CID 614367
IUPAC Name 2,7-ditert-butylnaphthalene
SMILES CC(C)(C)C1=CC2=C(C=C1)C=CC(=C2)C(C)(C)C

Specifications

Melting Point 104°C
Color White
Quantity 1 g
Formula Weight 240.39
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Di-tert-butylnaphthalene