accessibility menu, dialog, popup

UserName

H3CCH3H3CCH3H3CCH3

CAS RN 10275-58-8

IUPAC Name: 2,7-di-tert-butylnaphthalene
Synonyms: 2,7-di-tert-butylnaphthalene
Molecular Weight (g/mol): 240.39
Molecular Formula: C18H24
InChi Key: IHRUNSCACKKOPP-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=CC(=CC=C2C=C1)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results