missing translation for 'onlineSavingsMsg'
Learn More

2-[(7-Bromo-2,1,3-benzothiadiazol-4-yl)methylene]malononitrile 98.0+%, TCI America™

Catalog No. B5474200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5474200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5474200MG Supplier TCI America Supplier No. B5474200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1335150-10-1
Molecular Formula C10H3BrN4S
Molecular Weight (g/mol) 291.126
MDL Number MFCD23704429
InChI Key ZNWYPDKJWGLSQU-UHFFFAOYSA-N
PubChem CID 89394801
IUPAC Name 2-[(4-bromo-2,1,3-benzothiadiazol-7-yl)methylidene]propanedinitrile
SMILES C1=C(C2=NSN=C2C(=C1)Br)C=C(C#N)C#N

Specifications

Melting Point 189°C
Color Yellow
Quantity 200 mg
Formula Weight 291.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-[(7-Bromo-2,1,3-benzothiadiazol-4-yl)methylene]malononitrile
TSCA No
Recommended Storage Freezer