accessibility menu, dialog, popup

UserName

CAS RN 1335150-10-1

IUPAC Name: 2-[(7-bromo-2,1,3-benzothiadiazol-4-yl)methylidene]propanedinitrile
Synonyms: 2-[(7-bromo-2,1,3-benzothiadiazol-4-yl)methylidene]propanedinitrile
Molecular Weight (g/mol): 291.13
Molecular Formula: C10H3BrN4S
InChi Key: ZNWYPDKJWGLSQU-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C(C#N)C#N)C2=NSN=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results