missing translation for 'onlineSavingsMsg'
Learn More

2,7-Bis[N,N-bis(4-methoxyphenyl)amino]-9,9-spirobi[9H-fluorene] 98.0+%, TCI America™

Catalog No. p-7123866
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4959200MG 200 mg
B49591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4959200MG Supplier TCI America Supplier No. B4959200MG

Chemical Identifiers

CAS 1138220-69-5
Molecular Formula C53H42N2O4
Molecular Weight (g/mol) 770.929
InChI Key LZHVTCXAXYYCIF-UHFFFAOYSA-N
Synonym N,N,N′C,N′C-Tetrakis(4-methoxyphenyl)-9,9′C-spirobi[9H-fluorene]-2,7-diamine
PubChem CID 66616716
IUPAC Name 2-N',2-N',7-N',7-N'-tetrakis(4-methoxyphenyl)-9,9'-spirobi[fluorene]-2',7'-diamine
SMILES COC1=CC=C(C=C1)N(C2=CC=C(C=C2)OC)C3=CC4=C(C=C3)C5=C(C46C7=CC=CC=C7C8=CC=CC=C68)C=C(C=C5)N(C9=CC=C(C=C9)OC)C1=CC=C(C=C1)OC

Specifications

Melting Point 186°C
Color Yellow
Quantity 200 mg
Formula Weight 770.93
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis[N,N-bis(4-methoxyphenyl)amino]-9,9-spirobi[9H-fluorene]