accessibility menu, dialog, popup

UserName

CAS RN 1138220-69-5

IUPAC Name: N2,N2,N7,N7-tetrakis(4-methoxyphenyl)-9,9'-spirobi[fluorene]-2,7-diamine
Synonyms: N2,N2,N7,N7-tetrakis(4-methoxyphenyl)-9,9'-spirobi[fluorene]-2,7-diamine
Molecular Weight (g/mol): 770.93
Molecular Formula: C53H42N2O4
InChi Key: LZHVTCXAXYYCIF-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N(C1=CC=C(OC)C=C1)C1=CC2=C(C=C1)C1=C(C=C(C=C1)N(C1=CC=C(OC)C=C1)C1=CC=C(OC)C=C1)C21C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results