missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Bis(9H-carbazol-9-yl)-9,9-dimethylfluorene 98.0+%, TCI America™
Supplier: TCI America B49601G
Specifications
| 2,7-Bis(9H-carbazol-9-yl)-9,9-dimethylfluorene | |
| 286°C | |
| C39H28N2 | |
| 1 g | |
| IEQGNDONCZPWMW-UHFFFAOYSA-N | |
| 9-(7-carbazol-9-yl-9,9-dimethylfluoren-2-yl)carbazole | |
| 53420405 | |
| ≥98.0% (HPLC,N) |
| 226958-06-1 | |
| White-Yellow | |
| MFCD12022460 | |
| 9,9′C-(9,9-Dimethyl-9H-fluorene-2,7-diyl)bis(9H-carbazole), DMFL-CBP | |
| CC1(C2=C(C=CC(=C2)N3C4=CC=CC=C4C5=CC=CC=C53)C6=C1C=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97)C | |
| 524.667 | |
| 524.67 | |
| Crystalline Powder |
Chemical Identifiers
| 226958-06-1 | |
| 524.667 | |
| IEQGNDONCZPWMW-UHFFFAOYSA-N | |
| 53420405 | |
| CC1(C2=C(C=CC(=C2)N3C4=CC=CC=C4C5=CC=CC=C53)C6=C1C=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97)C |
| C39H28N2 | |
| MFCD12022460 | |
| 9,9′C-(9,9-Dimethyl-9H-fluorene-2,7-diyl)bis(9H-carbazole), DMFL-CBP | |
| 9-(7-carbazol-9-yl-9,9-dimethylfluoren-2-yl)carbazole |
Safety and Handling
TSCA : No