missing translation for 'onlineSavingsMsg'
Learn More

2,7-Bis(9H-carbazol-9-yl)-9,9-dimethylfluorene 98.0+%, TCI America™

Catalog No. p-7123867
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4960200MG 200 mg
B49601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4960200MG Supplier TCI America Supplier No. B4960200MG

Chemical Identifiers

CAS 226958-06-1
Molecular Formula C39H28N2
Molecular Weight (g/mol) 524.667
MDL Number MFCD12022460
InChI Key IEQGNDONCZPWMW-UHFFFAOYSA-N
Synonym 9,9′C-(9,9-Dimethyl-9H-fluorene-2,7-diyl)bis(9H-carbazole), DMFL-CBP
PubChem CID 53420405
IUPAC Name 9-(7-carbazol-9-yl-9,9-dimethylfluoren-2-yl)carbazole
SMILES CC1(C2=C(C=CC(=C2)N3C4=CC=CC=C4C5=CC=CC=C53)C6=C1C=C(C=C6)N7C8=CC=CC=C8C9=CC=CC=C97)C

Specifications

Melting Point 286°C
Color White-Yellow
Quantity 200 mg
Formula Weight 524.67
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis(9H-carbazol-9-yl)-9,9-dimethylfluorene