missing translation for 'onlineSavingsMsg'
Learn More

2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-dimethylfluorene 98.0+%, TCI America™

Catalog No. B51571G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51571G 1 g
B51575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51571G Supplier TCI America Supplier No. B51571G

Chemical Identifiers

CAS 325129-69-9
Molecular Formula C27H36B2O4
Molecular Weight (g/mol) 446.201
MDL Number MFCD08704230
InChI Key RVFLMSKITNJVRB-UHFFFAOYSA-N
Synonym 2,2'-9,9-dimethyl-9h-fluorene-2,7-diyl bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 9,9-dimethylfluorene-2,7-diboronic acid bis pinacol ester, 2-9,9-dimethyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl fluoren-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb256, 2,2-9,9-dimethyl-9h-fluorene-2,7-diyl, 9,9-dimethyl-9h-fluoren-2,7-diyl bisboronic acid pinacol ester, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-9,9-dimethylfluorene, 2,7-bis 4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl-9,9-dimethyl-9h-fluorene
PubChem CID 20640927
IUPAC Name 2-[9,9-dimethyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)fluoren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(C3(C)C)C=C(C=C4)B5OC(C(O5)(C)C)(C)C

Specifications

Color White
Quantity 1 g
Formula Weight 446.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9-dimethylfluorene