Learn More
2,6-Pyridinedicarboxylic acid chloride, 97%
CAS: 3739-94-4 | C7H3Cl2NO2 | 204.01 g/mol
$153.60 - $153.60
Chemical Identifiers
| CAS | 3739-94-4 |
|---|---|
| Molecular Formula | C7H3Cl2NO2 |
| Molecular Weight (g/mol) | 204.01 |
| MDL Number | MFCD00006289 |
| InChI Key | GWHOGODUVLQCEB-UHFFFAOYSA-N |
| Synonym | 2,6-pyridinedicarbonyl dichloride, 2,6-pyridinedicarboxylic acid chloride, pyridine-2,6-dicarbonyl dichloride, 2,6-pyridinedicarbonyl chloride, pyridine-2,6-dicarboxylic acid chloride, 2,6-pyridinedicarboxylicacidchloride, pyridine-2,6-dicarboyl dichloride, 2,6-pyridinedicarbony chloride, pubchem13243, tpc-i153 |
| PubChem CID | 77335 |
| IUPAC Name | pyridine-2,6-dicarbonyl chloride |
| SMILES | C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC162130050
|
Thermo Scientific Chemicals
162130050 |
5 g | Glass bottle |
Each for $153.60
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3739-94-4 | |
| 204.01 | |
| GWHOGODUVLQCEB-UHFFFAOYSA-N | |
| 77335 | |
| C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl |
| C7H3Cl2NO2 | |
| MFCD00006289 | |
| 2,6-pyridinedicarbonyl dichloride, 2,6-pyridinedicarboxylic acid chloride, pyridine-2,6-dicarbonyl dichloride, 2,6-pyridinedicarbonyl chloride, pyridine-2,6-dicarboxylic acid chloride, 2,6-pyridinedicarboxylicacidchloride, pyridine-2,6-dicarboyl dichloride, 2,6-pyridinedicarbony chloride, pubchem13243, tpc-i153 | |
| pyridine-2,6-dicarbonyl chloride |
Specifications
| 3739-94-4 | |
| 100.0 | |
| White to Brown | |
| Authentic | |
| Glass bottle | |
| (C5H3N)(COCl)2 | |
| 5 g | |
| 2,6-pyridinedicarbonyl dichloride, 2,6-pyridinedicarboxylic acid chloride, pyridine-2,6-dicarbonyl dichloride, 2,6-pyridinedicarbonyl chloride, pyridine-2,6-dicarboxylic acid chloride, 2,6-pyridinedicarboxylicacidchloride, pyridine-2,6-dicarboyl dichloride, 2,6-pyridinedicarbony chloride, pubchem13243, tpc-i153 | |
| C1=CC(=NC(=C1)C(=O)Cl)C(=O)Cl | |
| 204.01 | |
| 204.01 | |
| Crystalline Powder, Crystals or Chunks |
| 96.0 | |
| 57.0°C to 61.0°C | |
| 284.0°C | |
| 97% | |
| C7H3Cl2NO2 | |
| MFCD00006289 | |
| 22, 155 | |
| GWHOGODUVLQCEB-UHFFFAOYSA-N | |
| pyridine-2,6-dicarbonyl chloride | |
| 77335 | |
| 97% | |
| 2, 6-Pyridinedicarboxylic acid chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 223-125-4
RUO – Research Use Only