missing translation for 'onlineSavingsMsg'
Learn More

2-(6-Methoxynaphthalen-2-yl)acetic acid Pharmaceutical Impurity Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013659
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013659 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013659 Supplier Merck Emd Millipore Supplier No. 0813750MG
Only null left
Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 23981-47-7
Linear Formula C13H12O3
MDL Number MFCD00033105
Synonym Naproxen impurity I; 6-Methoxy-2-naphthaleneacetic acid; 6-Methoxy-2-naphthylacetic acid
Type Pharmaceutical Impurity Standard
Quantity 50 mg
Formula Weight 216.23
Percent Purity ≥98.0% (HPLC)