missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diphenylphenol 98.0+%, TCI America™

Catalog No. D25635G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25635G 5 g
D256325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D25635G Supplier TCI America Supplier No. D25635G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2432-11-3
Molecular Formula C18H14O
Molecular Weight (g/mol) 246.31
MDL Number MFCD00009716
InChI Key ATGFTMUSEPZNJD-UHFFFAOYSA-N
Synonym 1,1':3',1-terphenyl-2'-ol, m-terphenyl-2'-ol, 2,6-diphenyl phenol, 2'-hydroxy-m-terphenyl, unii-la93w35m96, 3-phenyl-1,1'-biphenyl-2-ol, 2,6-di-phenylphenol, 2,6,-diphenylphenol, 2,6-di phenyl phenol
PubChem CID 75512
IUPAC Name 2,6-diphenylphenol
SMILES C1=CC=C(C=C1)C2=C(C(=CC=C2)C3=CC=CC=C3)O

Specifications

Melting Point 103°C
Color White-Yellow
Quantity 5 g
Formula Weight 246.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diphenylphenol